2,3-Dimethylpyrazine ≥98.0% (by GC)

Supplier: TCI America

D1525-25ML D1525-5ML
TCD1525-25MLEA 37 USD
TCD1525-25ML TCD1525-5ML
2,3-Dimethylpyrazine ≥98.0% (by GC)
2,3-Dimethylpyrazine

CAS Number: 5910-89-4
MDL Number: MFCD00006144
Molecular Formula: C6H8N2
Molecular Weight: 108.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 156
Flash Point (°C): 57
Specific Gravity (20/20): 1.01

Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 156 °C
Density: 1.01 (20 °C)
Flash Pt: 57 °C
MDL Number: MFCD00006144
CAS Number: 5910-89-4
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2aaab9236fb72588a9a4944248b5846b20ac9cdc&vid=2aaab9236fb72588a9a4944248b5846b20ac9cdc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR