Dabsyl chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: DABS-Cl, 4-Dimethylaminoazobenzene-4'-sulphonyl Chloride, 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonyl chloride

D1382-1G D1382-25G
TCD1382-1GEA 130.17 USD
TCD1382-1G TCD1382-25G
Dabsyl chloride ≥98.0% (by titrimetric analysis)
Dabsyl chloride

[N-Protecting Agent for Peptides Research]
CAS Number: 56512-49-3
MDL Number: MFCD00007444
Molecular Formula: C14H14ClN3O2S
Molecular Weight: 323.80
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Analytical Science, UV Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: 4-Dimethylaminoazobenzene-4'-sulfonyl Chloride

Formula: C₁₄H₁₄ClN₃O₂S
MW: 323.80 g/mol
MDL Number: MFCD00007444
CAS Number: 56512-49-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa90a9f12891c1cfb1d0060b580730a92b55851d&vid=fa90a9f12891c1cfb1d0060b580730a92b55851d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR