N,N-Dimethylformamide di-tert-butyl acetal ≥98.0%

Supplier: TCI America

Synonyms: 1,1-Bis(1,1-dimethylethoxy)-N,N,N-trimethylamine, 1,1-Di-tert-butoxy-N,N-dimethylmethanamine, 1,1-Di-tert-butoxytrimethylamine

D1303-25ML D1303-5ML
TCD1303-25MLEA 325.4 USD
TCD1303-25ML TCD1303-5ML
N,N-Dimethylformamide di-tert-butyl acetal ≥98.0%
N,N-Dimethylformamide di-tert-butyl acetal

[for Esterification]
CAS Number: 36805-97-7
MDL Number: MFCD00015002
Molecular Formula: C11H25NO2
Molecular Weight: 203.33
Purity/Analysis Method: >98.0% (N)
Form: Clear Liquid
Boiling point (°C): 57
Flash Point (°C): 33
Specific Gravity (20/20): 0.85


Product Categories: Synthetic Organic Chemistry, Alkylation (Esterification), Analytical Science, Esterification & Alkylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: 1,1-Di-tert-butoxytrimethylamine

Formula: C₁₁H₂₅NO₂
Boiling Pt: 57 °C
Density: 0.85 (20 °C)
Flash Pt: 33 °C
MDL Number: MFCD00015002
CAS Number: 36805-97-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ff18009e98f842ee2c06555166a532463408952&vid=f45857cedac8441de1e2c3c05f6ea742ec667397&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR