N,N-Dimethylformamide dipropyl acetal ≥90.0%

Supplier: TCI America

Synonyms: 1,1-Dipropoxytrimethylamine, N,n-dimethylformamidedipropylacetal, 1,1-Dipropoxy-N,N-dimethylmethylamine

D1301-25ML D1301-5ML
TCD1301-25MLEA 84.7 USD
TCD1301-25ML TCD1301-5ML
N,N-Dimethylformamide dipropyl acetal ≥90.0%
N,N-Dimethylformamide dipropyl acetal

[for Esterification]
CAS Number: 6006-65-1
MDL Number: MFCD00009374
Molecular Formula: C9H21NO2
Molecular Weight: 175.27
Purity/Analysis Method: >90.0% (T)
Form: Clear Liquid
Flash Point (°C): 46
Specific Gravity (20/20): 0.85


Product Categories: Synthetic Organic Chemistry, Alkylation (Esterification), Analytical Science, Esterification & Alkylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: 1,1-Dipropoxytrimethylamine

Formula: C₉H₂₁NO₂
Density: 0.85 (20 °C)
Flash Pt: 46 °C
MDL Number: MFCD00009374
CAS Number: 6006-65-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebb7c90eb931a630ca5240617e4c8e910e2202a8&vid=07029c25c3533e7ee65f96860e27497a378cff87&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR