N,N-Dimethylformamide diethyl acetal ≥95.0%

Supplier: TCI America

Synonyms: 1,1-Diethoxy-N,N-dimethylmethanamine, 1,1-Diethoxytrimethylamine

D1294-25ML D1294-5ML
TCD1294-25MLEA 60 USD
TCD1294-25ML TCD1294-5ML
N,N-Dimethylformamide diethyl acetal ≥95.0%
N,N-Dimethylformamide diethyl acetal

[for Esterification]
CAS Number: 1188-33-6
MDL Number: MFCD00009227
Molecular Formula: C7H17NO2
Molecular Weight: 147.22
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 22
Specific Gravity (20/20): 0.86


Product Categories: Synthetic Organic Chemistry, Alkylation (Esterification), Analytical Science, Esterification & Alkylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: 1,1-Diethoxytrimethylamine

Formula: C₇H₁₇NO₂
Boiling Pt: 130 °C
Density: 0.86 (20 °C)
Flash Pt: 22 °C
MDL Number: MFCD00009227
CAS Number: 1188-33-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae855a2459b1086c9ec0653e39cf0f00019f543b&vid=1d20b376e4ac78f98748593a221840fa95608c95&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR