2,6-Dimethylcyclohexanone ≥98.0% (by GC)

Supplier: TCI America

D1280-250ML D1280-25ML
TCD1280-250MLEA 534 USD
TCD1280-250ML TCD1280-25ML
2,6-Dimethylcyclohexanone ≥98.0% (by GC)
2,6-Dimethylcyclohexanone

CAS Number: 2816-57-1
MDL Number: MFCD00001637
Molecular Formula: C8H14O
Molecular Weight: 126.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 51
Specific Gravity (20/20): 0.91

Formula: C₈H₁₄O
MW: 126.2 g/mol
Density: 0.91 (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD00001637
CAS Number: 2816-57-1
UN: 1224
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f81090d235c37179c950b054672e44ccc76cf6a1&vid=c4f80789f2a843560f3dc94acbb52605cea5c26d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR