2,4-Dibromoaniline ≥98.0% (by GC)

Supplier: TCI America

D1153-10G D1153-25G
TCD1153-10GEA 47 USD
TCD1153-10G TCD1153-25G
2,4-Dibromoaniline ≥98.0% (by GC)
2,4-Dibromoaniline

CAS Number: 615-57-6
MDL Number: MFCD00007633
Molecular Formula: C6H5Br2N
Molecular Weight: 250.92
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 156
Melting point (°C): 80

Formula: C₆H₅Br₂N
MW: 250.92 g/mol
Boiling Pt: 156 °C
Melting Pt: 80 °C
MDL Number: MFCD00007633
CAS Number: 615-57-6
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31824ce5012d5154f049cdae5779997716ec4139&vid=2fd3e5bc1819cded3b48807d487fa0e7645180e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR