3,4-Diaminopyridine ≥99.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3,4-Pyridinediamine, Amifampridine, 3,4-DAP, Pyridine-3,4-diyldiamine, Pyridine-3,4-diamine

D1149-1G D1149-5G
TCD1149-1GEA 36 USD
TCD1149-1G TCD1149-5G
3,4-Diaminopyridine ≥99.0% (by GC, titration analysis)
3,4-Diaminopyridine

CAS Number: 54-96-6
MDL Number: MFCD00006401
Molecular Formula: C5H7N3
Molecular Weight: 109.13
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 220
Flash Point (°C): 240

Formula: C₅H₇N₃
MW: 109.13 g/mol
Melting Pt: 220 °C
Flash Pt: 240 °C
MDL Number: MFCD00006401
CAS Number: 54-96-6
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b878e1d9820f89b07f0bea437534c3ac730713c&vid=1b878e1d9820f89b07f0bea437534c3ac730713c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR