Dimethoxy(dimethyl)silane ≥98.0% (by GC)

Supplier: TCI America

D1052-25ML D1052-500ML
TCD1052-25MLEA 25.43 USD
TCD1052-25ML TCD1052-500ML
Dimethoxy(dimethyl)silane ≥98.0% (by GC)
Dimethoxy(dimethyl)silane

CAS Number: 1112-39-6
MDL Number: MFCD00025691
Molecular Formula: C4H12O2Si
Molecular Weight: 120.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 81
Flash Point (°C): -5
Specific Gravity (20/20): 0.87

Formula: C₄H₁₂O₂Si
Boiling Pt: 81 °C
Density: 0.87 (20 °C)
Flash Pt: –5 °C
MDL Number: MFCD00025691
CAS Number: 1112-39-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=065a43073b72eef6705bc23846e897d2eb01d55f&vid=065a43073b72eef6705bc23846e897d2eb01d55f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR