Dodecylamine ≥97.0% (by GC, titration analysis)
Supplier: TCI America
CAS Number: 124-22-1
MDL Number: MFCD00008154
Molecular Formula: C12H27N
Molecular Weight: 185.36
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 259
Flash Point (°C): 178
Freezing point (°C): 26
Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 1-Aminododecane, Laurylamine
Formula:
H₃C(CH₂)₁₀CH₂NH₂ Boiling Pt: 259 °C Flash Pt: 178 °C |
MDL Number:
MFCD00008154 CAS Number: 124-22-1 UN: 3259 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...