2,3-Dimethylbutane ≥98.0% (by GC)

Supplier: TCI America

D0690-25ML D0690-500ML D0690-5ML
TCD0690-25MLEA 49 USD
TCD0690-25ML TCD0690-500ML TCD0690-5ML
2,3-Dimethylbutane ≥98.0% (by GC)
2,3-Dimethylbutane

CAS Number: 79-29-8
MDL Number: MFCD00008925
Molecular Formula: C6H14
Molecular Weight: 86.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 58
Flash Point (°C): -29
Specific Gravity (20/20): 0.66


Other Product Names / Synonyms: Diisopropyl

Formula: C₆H₁₄
Boiling Pt: 58 °C
Density: 0.66 (20 °C)
Flash Pt: –29 °C
MDL Number: MFCD00008925
CAS Number: 79-29-8
UN: 2457
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e12b41740ff1d82b6163cc483391e925e66df94&vid=efadd8081432b6c89ca1ded9e8a24e9cbd59da40&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR