Dansyl chloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
5-(Dimethylamino)naphthalene-1-sulphonyl chloride
CAS Number: 605-65-2
MDL Number: MFCD00003985
Molecular Formula: C12H12ClNO2S
Molecular Weight: 269.74
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 70
Storage Temperature: 0-10°C
Product Categories: Bioscience, Fluorescent Labeling Dyes, Analytical Science, Derivatization Reagents for MS, Fluorescence Detection (HPLC Labeling Reagents), Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: 5-(Dimethylamino)naphthalene-1-sulfonyl Chloride
Formula:
C₁₂H₁₂ClNO₂S MW: 269.75 g/mol Melting Pt: 70 °C |
MDL Number:
MFCD00003985 CAS Number: 605-65-2 UN: 3261 ADR: 8,III |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...