1,4-Diethylbenzene ≥98.0% (by GC)

Supplier: TCI America

D0479-25ML D0479-5ML
TCD0479-25MLEA 144.4 USD
TCD0479-25ML TCD0479-5ML
1,4-Diethylbenzene ≥98.0% (by GC)
1,4-Diethylbenzene

CAS Number: 105-05-5
MDL Number: MFCD00009264
Molecular Formula: C10H14
Molecular Weight: 134.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 184
Melting point (°C): -43
Flash Point (°C): 55
Specific Gravity (20/20): 0.86

Formula: C₁₀H₁₄
MW: 134.22 g/mol
Boiling Pt: 184 °C
Melting Pt: -43 °C
Density: 0.86 (20 °C)
Flash Pt: 55 °C
MDL Number: MFCD00009264
CAS Number: 105-05-5
UN: 2049
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7141f0a1235b2239e8d7714ce9054b42ccb4f59e&vid=7141f0a1235b2239e8d7714ce9054b42ccb4f59e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR