trans-1,4-Dimethylcyclohexane ≥95.0% (by GC)

Supplier: TCI America

Synonyms: trans-Hexahydro-p-xylene

D0271-25ML D0271-5ML
TCD0271-25MLEA 258 USD
TCD0271-25ML TCD0271-5ML
trans-1,4-Dimethylcyclohexane ≥95.0% (by GC)
trans-1,4-Dimethylcyclohexane

CAS Number: 2207-04-7
MDL Number: MFCD00064955
Molecular Formula: C8H16
Molecular Weight: 112.22
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 119
Flash Point (°C): 6
Specific Gravity (20/20): 0.76


Other Product Names / Synonyms: trans-Hexahydro-p-xylene

Formula: C₈H₁₆
MW: 112.22 g/mol
Boiling Pt: 119 °C
Density: 0.76 (20 °C)
Flash Pt: 6 °C
MDL Number: MFCD00064955
CAS Number: 2207-04-7
UN: 2263
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ee8ae029e0c2af865a16c815d2c4ade069e29b0&vid=5ee8ae029e0c2af865a16c815d2c4ade069e29b0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR