Diisopropylcarbodiimide ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
N,N'-methanetetraylbis(1-methylethylamine), N,N-Diisopropylcarbodiimide, 1,3-Diisopropylcarbodiimide, N,N'-Diisopropylcarbodiimide, N.N’-Diisopropylcarbodiimide, DIC
CAS Number: 693-13-0
MDL Number: MFCD00065689
Molecular Formula: C7H14N2
Molecular Weight: 126.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 147
Flash Point (°C): 31
Specific Gravity (20/20): 0.82
Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Peptide Synthesis, Bioscience, Crosslinkers
Other Product Names / Synonyms: DIC
Formula:
C₇H₁₄N₂ MW: 126.2 g/mol Boiling Pt: 147 °C Density: 0.82 (20 °C) Flash Pt: 31 °C |
MDL Number:
MFCD00065689 CAS Number: 693-13-0 UN: 2929 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...