2,4-Dinitrophenol ≥98.0% (by GC and titration analysis on dried basis) wetted with ca. 20% water

Supplier: TCI America

Synonyms: 1,3-Dinitro-4-hydroxybenzene, α-Dinitrophenol

D0109-25G D0109-300G
TCD0109-25GEA 48.76 USD
TCD0109-25G TCD0109-300G
2,4-Dinitrophenol ≥98.0% (by GC and titration analysis on dried basis) wetted with ca. 20% water
2,4-Dinitrophenol

CAS Number: 51-28-5
MDL Number: MFCD00007115
Molecular Formula: C6H4N2O5
Molecular Weight: 184.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 113

Formula: C₆H₄N₂O₅
MW: 184.11 g/mol
Melting Pt: 113 °C
CAS Number: 51-28-5
UN: 1320
ADR: 4.1,I

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR