1,7-Diaminoheptane ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Heptamethylendiamine, Heptane-1,7-diamine, 1,7-Heptanediamine

D0094-25G D0094-5G
TCD0094-25GEA 156 USD
TCD0094-25G TCD0094-5G
1,7-Diaminoheptane ≥98.0% (by GC, titration analysis)
1,7-Diaminoheptane

CAS Number: 646-19-5
MDL Number: MFCD00008246
Molecular Formula: C7H18N2
Molecular Weight: 130.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 225
Flash Point (°C): 88
Freezing point (°C): 27


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Heptamethylenediamine, 1,7-Heptanediamine

Formula: C₇H₁₈N₂
MW: 130.23 g/mol
Boiling Pt: 225 °C
Flash Pt: 88 °C
MDL Number: MFCD00008246
CAS Number: 646-19-5
UN: 3295
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8d78210fd3293bc2f9f8e7ff80cf8ad0e44332c&vid=d8d78210fd3293bc2f9f8e7ff80cf8ad0e44332c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR