Decanoyl chloride ≥98.0%

Supplier: TCI America

Synonyms: Decanoylchloride

D0027-25ML D0027-500ML
TCD0027-25MLEA 17.29 USD
TCD0027-25ML TCD0027-500ML
Decanoyl chloride ≥98.0%
Decanoyl chloride

CAS Number: 112-13-0
MDL Number: MFCD00000771
Molecular Formula: C10H19ClO
Molecular Weight: 190.71
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 230
Flash Point (°C): 106
Specific Gravity (20/20): 0.94

Formula: C₁₀H₁₉ClO
Boiling Pt: 230 °C
Density: 0.94 (20 °C)
Flash Pt: 106 °C
MDL Number: MFCD00000771
CAS Number: 112-13-0
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=955c7f44338c42d05771586628c651992bae820e&vid=955c7f44338c42d05771586628c651992bae820e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR