Clonixin ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
2-[(3-Chloro-2-methylphenyl)amino]nicotinic Acid, 2-(3-Chloro-2-methylanilino)pyridine-3-carboxylic acid, 2-(3-Chloro-o-toluidino)nicotinic Acid
CAS Number: 17737-65-4
MDL Number: MFCD00864292
Molecular Formula: C13H11ClN2O2
Molecular Weight: 262.69
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 242
Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Anti-rheumatoid Arthritis Ingredients, Anti-inflammatory Ingredients, Anti-inflammatory Ingredients
Other Product Names / Synonyms: 2-(3-Chloro-o-toluidino)nicotinic Acid, 2-[(3-Chloro-2-methylphenyl)amino]nicotinic Acid
Formula:
C₁₃H₁₁ClN₂O₂ MW: 262.69 g/mol Melting Pt: 242 °C |
MDL Number:
MFCD00864292 CAS Number: 17737-65-4 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...