2-Chloro-5-fluoropyrimidine ≥98.0% (by GC)

Supplier: TCI America

C2546-1G C2546-5G
TCC2546-1GEA 49 USD
TCC2546-1G TCC2546-5G
2-Chloro-5-fluoropyrimidine ≥98.0% (by GC)
2-Chloro-5-fluoropyrimidine

CAS Number: 62802-42-0
MDL Number: MFCD03788197
Molecular Formula: C4H2ClFN2
Molecular Weight: 132.52
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 94
Flash Point (°C): 65
Specific Gravity (20/20): 1.44

Formula: C₄H₂ClFN₂
MW: 132.52 g/mol
Boiling Pt: 94 °C
Density: 1.44 (20 °C)
Flash Pt: 65 °C
MDL Number: MFCD03788197
CAS Number: 62802-42-0
UN: 3267
ADR: 8,III

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR