3-Chloro-2-fluoropyridine ≥98.0% (by GC)
Supplier: TCI America
C2515-25G
C2515-5G
TCC2515-25GEA
258.4
USD
TCC2515-25G
TCC2515-5G
3-Chloro-2-fluoropyridine ≥98.0% (by GC)
3-Chloro-2-fluoropyridine
CAS Number: 1480-64-4
MDL Number: MFCD04114144
Molecular Formula: C5H3ClFN
Molecular Weight: 131.53
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 70
Flash Point (°C): 52
Specific Gravity (20/20): 1.34
Formula:
C₅H₃ClFN Boiling Pt: 70 °C Density: 1.34 (20 °C) Flash Pt: 52 °C |
MDL Number:
MFCD04114144 CAS Number: 1480-64-4 UN: 1992 ADR: 3,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...