α-Bromo-4-chloro-2-fluorotoluene ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Chloro-2-fluorobenzyl bromide, 1-(Bromomethyl)-4-chloro-2-fluorobenzene

C2483-25G C2483-5G
TCC2483-25GEA 230 USD
TCC2483-25G TCC2483-5G
α-Bromo-4-chloro-2-fluorotoluene ≥98.0% (by GC, titration analysis)
α-Bromo-4-chloro-2-fluorotoluene

CAS Number: 71916-82-0
MDL Number: MFCD00831105
Molecular Formula: C7H5BrClF
Molecular Weight: 223.47
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 98
Specific Gravity (20/20): 1.65
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1-(Bromomethyl)-4-chloro-2-fluorobenzene

Formula: C₇H₅BrClF
MW: 223.47 g/mol
Boiling Pt: 98 °C
Density: 1.65 (20 °C)
MDL Number: MFCD00831105
CAS Number: 71916-82-0
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28026d937d72f682813219bfa3961e0e7d7871bd&vid=28026d937d72f682813219bfa3961e0e7d7871bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR