α-Bromo-3-chloro-4-fluorotoluene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3-Chloro-4-fluorobenzyl bromide, 4-(Bromomethyl)-2-chloro-1-fluorobenzene

C2482-25G C2482-5G
TCC2482-25GEA 137 USD
TCC2482-25G TCC2482-5G
α-Bromo-3-chloro-4-fluorotoluene ≥98.0% (by GC)
α-Bromo-3-chloro-4-fluorotoluene

CAS Number: 192702-01-5
MDL Number: MFCD01631551
Molecular Formula: C7H5BrClF
Molecular Weight: 223.47
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 65
Specific Gravity (20/20): 1.66


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha-Bromo-3-chloro-4-fluorotoluene, 1-(Bromomethyl)-3-chloro-4-fluorobenzene

Formula: C₇H₅BrClF
MW: 223.47 g/mol
Boiling Pt: 65 °C
Density: 1.66 (20 °C)
MDL Number: MFCD01631551
CAS Number: 192702-01-5
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=359857f51460df37facbc0e186bd80f6c48c4645&vid=359857f51460df37facbc0e186bd80f6c48c4645&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR