Chlorpromazine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
2-Chloro-10-[3-(dimethylamino)-1-propyl]phenothiazine Hydrochloride
C2481-25G
C2481-5G
TCC2481-25GEA
121.06
USD
TCC2481-25G
TCC2481-5G
Chlorpromazine hydrochloride ≥98.0% (by HPLC, titration analysis)
Chlorpromazine hydrochloride
CAS Number: 69-09-0
MDL Number: MFCD00012654
Molecular Formula: C17H19ClN2S
Molecular Weight: 355.32
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 197
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: 2-Chloro-10-[3-(dimethylamino)-1-propyl]phenothiazine Hydrochloride
Formula:
C₁₇H₁₉ClN₂S MW: 355.32 g/mol Melting Pt: 197 °C |
MDL Number:
MFCD00012654 CAS Number: 69-09-0 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...