α-Bromo-3-chloro-5-fluorotoluene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-(Bromomethyl)-3-chloro-5-fluorobenzene, 3-Chloro-5-fluorobenzyl bromide

C2474-1G C2474-5G
TCC2474-1GEA 45 USD
TCC2474-1G TCC2474-5G
α-Bromo-3-chloro-5-fluorotoluene ≥98.0% (by GC)
α-Bromo-3-chloro-5-fluorotoluene

CAS Number: 493024-39-8
MDL Number: MFCD03788472
Molecular Formula: C7H5BrClF
Molecular Weight: 223.47
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.65


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-(Bromomethyl)-3-chloro-5-fluorobenzene

Formula: C₇H₅BrClF
MW: 223.47 g/mol
Density: 1.65 (20 °C)
MDL Number: MFCD03788472
CAS Number: 493024-39-8
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cca0bac6806a7defa41e92f52129e7b39ac0534d&vid=cca0bac6806a7defa41e92f52129e7b39ac0534d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR