2-Chlorothiazole ≥98.0% (by GC)
Supplier: TCI America
C2471-25G
C2471-5G
TCC2471-25GEA
149.6
USD
TCC2471-25G
TCC2471-5G
2-Chlorothiazole ≥98.0% (by GC)
2-Chlorothiazole
CAS Number: 3034-52-4
MDL Number: MFCD00210701
Molecular Formula: C3H2ClNS
Molecular Weight: 119.57
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 145
Flash Point (°C): 54
Specific Gravity (20/20): 1.40
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 2-Thiazolyl Chloride
Formula:
C₃H₂ClNS MW: 119.57 g/mol Boiling Pt: 145 °C Density: 1.40 (20 °C) Flash Pt: 54 °C |
MDL Number:
MFCD00210701 CAS Number: 3034-52-4 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...