(Chloromethyl)cyclopropane ≥95.0% (by GC)

Supplier: TCI America

Synonyms: Cyclopropyl methyl chloride

C2469-25G C2469-5G
TCC2469-25GEA 118 USD
TCC2469-25G TCC2469-5G
(Chloromethyl)cyclopropane ≥95.0% (by GC)
(Chloromethyl)cyclopropane

CAS Number: 5911-08-0
MDL Number: MFCD00001307
Molecular Formula: C4H7Cl
Molecular Weight: 90.55
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Melting point (°C): -91
Flash Point (°C): -2
Specific Gravity (20/20): 0.99

Formula: C₄H₇Cl
MW: 90.55 g/mol
Boiling Pt: 88 °C
Melting Pt: -91 °C
Density: 0.99 (20 °C)
Flash Pt: -2 °C
MDL Number: MFCD00001307
CAS Number: 5911-08-0
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d8706ca3f338fce9d9781df29930822f8a52ebf&vid=2d8706ca3f338fce9d9781df29930822f8a52ebf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR