3-Chloropropionaldehyde dimethyl acetal ≥93.0%
Supplier: TCI America
Synonyms:
1-Chloro-3,3-dimethoxypropane
C2458-25G
C2458-5G
TCC2458-25GEA
313
USD
TCC2458-25G
TCC2458-5G
3-Chloropropionaldehyde dimethyl acetal ≥93.0%
3-Chloropropionaldehyde dimethyl acetal
CAS Number: 35502-06-8
MDL Number: MFCD16038653
Molecular Formula: C5H11ClO2
Molecular Weight: 138.59
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Boiling point (°C): 45
Specific Gravity (20/20): 1.06
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 1-Chloro-3,3-dimethoxypropane
Formula:
C₅H₁₁ClO₂ Boiling Pt: 45 °C Density: 1.06 (20 °C) |
MDL Number:
MFCD16038653 CAS Number: 35502-06-8 |
|
Specification Test Results
Purity/Analysis Method | >93.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...