Cyclopropanemethylamine ≥95.0%

Supplier: TCI America

Synonyms: Cyclopropylmethylamine, (Cyclopropylmethyl)amine, (Aminomethyl)cyclopropane

C2345-25G C2345-5G
TCC2345-25GEA 368 USD
TCC2345-25G TCC2345-5G
Cyclopropanemethylamine ≥95.0%
Cyclopropanemethylamine

CAS Number: 2516-47-4
MDL Number: MFCD00037147
Molecular Formula: C4H9N
Molecular Weight: 71.12
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 86
Flash Point (°C): 2
Specific Gravity (20/20): 0.83


Other Product Names / Synonyms: (Aminomethyl)cyclopropane

Formula: C₄H₉N
MW: 71.12 g/mol
Boiling Pt: 84
Density: 0.83 (20 °C)
Flash Pt: 2 °C
MDL Number: MFCD00037147
CAS Number: 2516-47-4
UN: 2733
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3605c5c8d1fa1d0925901a4c29b97d3be0f2a643&vid=3605c5c8d1fa1d0925901a4c29b97d3be0f2a643&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR