5-Chloro-2-thiophenesulfonyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 5-Chlorothiophene-2-sulphonyl chloride

C2343-25G C2343-5G
TCC2343-25GEA 237 USD
TCC2343-25G TCC2343-5G
5-Chloro-2-thiophenesulfonyl chloride ≥98.0% (by GC, titration analysis)
5-Chloro-2-thiophenesulphonyl chloride

CAS Number: 2766-74-7
MDL Number: MFCD00051667
Molecular Formula: C4H2Cl2O2S2
Molecular Weight: 217.08
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 66
Flash Point (°C): 110
Storage Temperature: 0-10°C

Formula: C₄H₂Cl₂O₂S₂
MW: 217.1 g/mol
Boiling Pt: 66 °C
Flash Pt: 110 °C
MDL Number: MFCD00051667
CAS Number: 2766-74-7
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8130180a17b687a541edafc1a13a77fa8e8062c&vid=c8130180a17b687a541edafc1a13a77fa8e8062c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR