3-Chloro-4-nitrotoluene ≥98.0%

Supplier: TCI America

Synonyms: 2-Chloro-4-methyl-1-nitrobenzene

C2277-25G C2277-5G
TCC2277-25GEA 79.5 USD
TCC2277-25G TCC2277-5G
3-Chloro-4-nitrotoluene ≥98.0%
3-Chloro-4-nitrotoluene

CAS Number: 38939-88-7
MDL Number: MFCD02683043
Molecular Formula: C7H6ClNO2
Molecular Weight: 171.58
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 101
Flash Point (°C): 110
Freezing point (°C): 25

Formula: C₇H₆ClNO₂
Boiling Pt: 101 °C
Flash Pt: 110 °C
MDL Number: MFCD02683043
CAS Number: 38939-88-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea1476b4ce9f52870c8c0ba254894527bd134f70&vid=7816835d623c8c8f45b9833a7f5391bcaf98ef5c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR