Cyclopropylboronic acid pinacol ester ≥95.0% (by GC)
Supplier: TCI America
Synonyms:
2-Cyclopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
C2276-1G
C2276-5G
TCC2276-1GEA
136
USD
TCC2276-1G
TCC2276-5G
Cyclopropylboronic acid pinacol ester ≥95.0% (by GC)
Cyclopropylboronic acid pinacol ester
CAS Number: 126689-01-8
MDL Number: MFCD05663847
Molecular Formula: C9H17BO2
Molecular Weight: 168.04
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 146
Flash Point (°C): 40
Specific Gravity (20/20): 0.93
Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: (4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)cyclopropane, Cyclopropylboronic Acid Pinacol Ester
Formula:
C₉H₁₇BO₂ MW: 168.04 g/mol Boiling Pt: 146 °C Density: 0.93 (20 °C) Flash Pt: 40 °C |
MDL Number:
MFCD05663847 CAS Number: 126689-01-8 UN: 3272 ADR: 3,III |
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...