2-Chloro-5-fluoropyridine ≥98.0% (by GC)

Supplier: TCI America

C2270-1G C2270-5G
TCC2270-1GEA 24 USD
TCC2270-1G TCC2270-5G
2-Chloro-5-fluoropyridine ≥98.0% (by GC)
2-Chloro-5-fluoropyridine

CAS Number: 31301-51-6
MDL Number: MFCD03095332
Molecular Formula: C5H3ClFN
Molecular Weight: 131.53
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 74
Flash Point (°C): 42
Freezing point (°C): 19
Specific Gravity (20/20): 1.34

Formula: C₅H₃ClFN
MW: 131.54 g/mol
Boiling Pt: 74 °C
Density: 1.34 (20 °C)
Flash Pt: 42 °C
MDL Number: MFCD03095332
CAS Number: 31301-51-6
UN: 2929
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc087f29f59e950760d0e6a9913e1896ad7f65ed&vid=dc087f29f59e950760d0e6a9913e1896ad7f65ed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR