Carvedilol ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
1-(9H-Carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]-2-propanol
C2260-25G
C2260-5G
TCC2260-25GEA
633
USD
TCC2260-25G
TCC2260-5G
Carvedilol ≥98.0% (by HPLC, titration analysis)
Carvedilol
CAS Number: 72956-09-3
MDL Number: MFCD00864692
Molecular Formula: C24H26N2O4
Molecular Weight: 406.48
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 117
Product Categories: Bioscience, Antihypertensive Ingredients
Other Product Names / Synonyms: 1-(9H-Carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]-2-propanol
Formula:
C₂₄H₂₆N₂O₄ MW: 406.48 g/mol Melting Pt: 117 °C |
MDL Number:
MFCD00864692 CAS Number: 72956-09-3 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...