4-Chloro-3-(trifluoromethyl)benzonitrile ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Chloro-5-cyanobenzotrifluoride

C2246-25G C2246-5G
TCC2246-25GEA 192 USD
TCC2246-25G TCC2246-5G
4-Chloro-3-(trifluoromethyl)benzonitrile ≥98.0% (by GC)
4-Chloro-3-(trifluoromethyl)benzonitrile

CAS Number: 1735-54-2
MDL Number: MFCD00190117
Molecular Formula: C8H3ClF3N
Molecular Weight: 205.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 105
Melting point (°C): 65


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Chloro-5-cyanobenzotrifluoride

Formula: C₈H₃ClF₃N
MW: 205.57
Boiling Pt: 105 °C
Melting Pt: 65 °C
MDL Number: MFCD00190117
CAS Number: 1735-54-2
UN: 3439
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8debb4e38c180f9533cd46f4c691abf6e3d9e55b&vid=8debb4e38c180f9533cd46f4c691abf6e3d9e55b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR