2-Chloro-6-fluorobenzoyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C2229-25G C2229-5G
TCC2229-25GEA 73 USD
TCC2229-25G TCC2229-5G
2-Chloro-6-fluorobenzoyl chloride ≥98.0% (by GC, titration analysis)
2-Chloro-6-fluorobenzoyl chloride

CAS Number: 79455-63-3
MDL Number: MFCD00178746
Molecular Formula: C7H3Cl2FO
Molecular Weight: 193.00
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 1.43

Formula: C₇H₃Cl₂FO
Density: 1.43 (20 °C)
MDL Number: MFCD00178746
CAS Number: 79455-63-3
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac7aaec4d7661e9ddc0575401a47b00e12a95177&vid=ac7aaec4d7661e9ddc0575401a47b00e12a95177&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR