2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphordiamidite ≥95.0% (by GC, titration analysis)
Supplier: TCI America
CAS Number: 102691-36-1
MDL Number: MFCD00012213
Molecular Formula: C15H32N3OP
Molecular Weight: 301.41
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 60
Specific Gravity (20/20): 0.95
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Phosphorylation, Nucleic Acid Synthesis
Other Product Names / Synonyms: Bis(diisopropylamino)(2-cyanoethoxy)phosphine
Formula:
C₁₅H₃₂N₃OP Density: 0.95 (20 °C) Flash Pt: 60 °C |
MDL Number:
MFCD00012213 CAS Number: 102691-36-1 |
|
Specification Test Results
Purity/Analysis Method | >95.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...