2-Chloro-5-iodobenzonitrile ≥98.0% (by GC)

Supplier: TCI America

C2212-1G C2212-5G
TCC2212-1GEA 28 USD
TCC2212-1G TCC2212-5G
2-Chloro-5-iodobenzonitrile ≥98.0% (by GC)
2-Chloro-5-iodobenzonitrile

CAS Number: 289039-29-8
MDL Number: MFCD00672973
Molecular Formula: C7H3ClIN
Molecular Weight: 263.46
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 117

Formula: C₇H₃ClIN
MW: 263.4629 g/mol
Melting Pt: 117 °C
MDL Number: MFCD00672973
CAS Number: 289039-29-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7964f41bc6c3a8ef9d0fde49510084e7e0c698a6&vid=7964f41bc6c3a8ef9d0fde49510084e7e0c698a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR