Cesium fluoride ≥99.0%

Supplier: TCI America

C2204-100G C2204-25G
TCC2204-100GEA 183 USD
TCC2204-100G TCC2204-25G
Cesium fluoride ≥99.0%
Cesium fluoride

CAS Number: 13400-13-0
MDL Number: MFCD00010960
Molecular Formula: CsF
Molecular Weight: 151.90
Form: Crystal
Color: White
Melting point (°C): 602

Formula: CsF
MW: 151.9
Boiling Pt: 1251
Melting Pt: 680
Density: 4.627
MDL Number: MFCD00010960
CAS Number: 13400-13-0
UN: 2923
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b15cf83309c604d6c73fa26991368ece8e4b3ab9&vid=b15cf83309c604d6c73fa26991368ece8e4b3ab9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR