4-Chloro-1,1-dimethoxybutane ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 4-Chlorobutyraldehyde dimethyl Acetal, 4-Chloro-1,1-dimethoxybutane

C2179-25G C2179-5G
TCC2179-25GEA 149 USD
TCC2179-25G TCC2179-5G
4-Chloro-1,1-dimethoxybutane ≥97.0% (by GC)
4-Chloro-1,1-dimethoxybutane

CAS Number: 29882-07-3
MDL Number: MFCD01632883
Molecular Formula: C6H13ClO2
Molecular Weight: 152.62
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 43
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: 4-Chloro-1,1-dimethoxybutane

Formula: C₆H₁₃ClO₂
MW: 152.62 g/mol
Boiling Pt: 120 °C
Density: 1.04 (20 °C)
Flash Pt: 43 °C
MDL Number: MFCD09032953
CAS Number: 29882-07-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=129b7f5291e936106d5396f2a7426bcf569a9a80&vid=129b7f5291e936106d5396f2a7426bcf569a9a80&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR