2-Chloro-4-fluorobenzonitrile ≥98.0% (by GC)

Supplier: TCI America

C2129-25G C2129-5G
TCC2129-25GEA 179.9 USD
TCC2129-25G TCC2129-5G
2-Chloro-4-fluorobenzonitrile ≥98.0% (by GC)
2-Chloro-4-fluorobenzonitrile

CAS Number: 60702-69-4
MDL Number: MFCD00042523
Molecular Formula: C7H3ClFN
Molecular Weight: 155.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 65

Formula: C₇H₃ClFN
MW: 155.56 g/mol
Melting Pt: 65 °C
MDL Number: MFCD00042523
CAS Number: 60702-69-4
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42afc2517ea6230bf5db7e0c5f798aa4b02e1107&vid=e3b23fb6be90aab2557bd2df5c16bfbdd5f0395b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR