Chloroacetic acid ≥99.0%
Supplier: TCI America
Synonyms:
Monochloroacetic acid
C2123-25G
C2123-500G
TCC2123-25GEA
37.67
USD
TCC2123-25G
TCC2123-500G
Chloroacetic acid ≥99.0%
Chloroacetic acid
CAS Number: 79-11-8
MDL Number: MFCD00002683
Molecular Formula: C2H3ClO2
Molecular Weight: 94.49
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 189
Melting point (°C): 65
Flash Point (°C): 126
Product Categories: Synthetic Organic Chemistry, Carboxylic Acids (Multi-component Reaction)
Other Product Names / Synonyms: Monochloroacetic Acid
Formula:
ClCH₂COOH Boiling Pt: 189 °C Melting Pt: 65 °C Flash Pt: 126 °C |
MDL Number:
MFCD00002683 CAS Number: 79-11-8 UN: 1751 ADR: 6.1,II |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...