α-Bromo-3-chloro-2-fluorotoluene ≥96.0%

Supplier: TCI America

Synonyms: 3-Chloro-2-fluorobenzyl bromide, 1-(Bromomethyl)-3-chloro-2-fluorobenzene

C2118-1G C2118-5G
TCC2118-1GEA 77.4 USD
TCC2118-1G TCC2118-5G
α-Bromo-3-chloro-2-fluorotoluene ≥96.0%
α-Bromo-3-chloro-2-fluorotoluene

CAS Number: 85070-47-9
MDL Number: MFCD00042503
Molecular Formula: C7H5BrClF
Molecular Weight: 223.47
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Boiling point (°C): 126
Freezing point (°C): 30


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-(Bromomethyl)-3-chloro-2-fluorobenzene

Formula: C₇H₅BrClF
Boiling Pt: 126 °C
MDL Number: MFCD00042503
CAS Number: 85070-47-9
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=928969ed53b5061c9e1c387fcb48e2648cd79344&vid=928969ed53b5061c9e1c387fcb48e2648cd79344&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR