5-Chloro-2,3-difluoropyridine ≥93.0% (by GC)

Supplier: TCI America

C2113-25G C2113-5G
TCC2113-25GEA 61 USD
TCC2113-25G TCC2113-5G
5-Chloro-2,3-difluoropyridine ≥93.0% (by GC)
5-Chloro-2,3-difluoropyridine

CAS Number: 89402-43-7
MDL Number: MFCD04039314
Molecular Formula: C5H2ClF2N
Molecular Weight: 149.52
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Boiling point (°C): 135
Specific Gravity (20/20): 1.44

Formula: C5H2ClF2N
MW: 149.5259 g/mol
Boiling Pt: 135 °C
Density: 1.44 (20 °C)
MDL Number: MFCD04039314
CAS Number: 89402-43-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4a154eb09dcceddeec236bf814e4f7ea22fe7c6&vid=b4a154eb09dcceddeec236bf814e4f7ea22fe7c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >93.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR