(R)-(+)-2-Chlorobutyric acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C2109-1G C2109-5G
TCC2109-1GEA 151 USD
TCC2109-1G TCC2109-5G
(R)-(+)-2-Chlorobutyric acid ≥98.0% (by titrimetric analysis)
(R)-(+)-2-Chlorobutyric acid

CAS Number: 54053-45-1
MDL Number: MFCD00067098
Molecular Formula: C4H7ClO2
Molecular Weight: 122.55
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 98
Specific Gravity (20/20): 1.19
Specific rotation [a]20/D: 17 deg (neat)

Formula: C₄H₇ClO₂
Boiling Pt: 98 °C
Density: 1.19 (20 °C)
MDL Number: MFCD00067098
CAS Number: 54053-45-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9a7fb6e61df6067a6c775afa342d52a1e96cdc2&vid=b9a7fb6e61df6067a6c775afa342d52a1e96cdc2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR