α-Bromo-2-chloro-4-fluorotoluene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-(Bromomethyl)-2-chloro-4-fluorobenzene, 2-Chloro-4-fluorobenzyl bromide

C2098-25G C2098-5G
TCC2098-25GEA 118 USD
TCC2098-25G TCC2098-5G
α-Bromo-2-chloro-4-fluorotoluene ≥98.0% (by GC)
α-Bromo-2-chloro-4-fluorotoluene

CAS Number: 45767-66-6
MDL Number: MFCD00236025
Molecular Formula: C7H5BrClF
Molecular Weight: 223.47
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 39
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-(Bromomethyl)-2-chloro-4-fluorobenzene

Formula: C₇H₅BrClF
MW: 223.47 g/mol
Melting Pt: 39 °C
MDL Number: MFCD00236025
CAS Number: 45767-66-6
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=506d84189b33a4574dc2651be652f51f7c0a4678&vid=506d84189b33a4574dc2651be652f51f7c0a4678&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR