Indole-6-carbonitrile ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 6-Cyano-1H-indole, 6-Cyanoindole, 1H-Indole-6-carbonitrile

C2084-25G C2084-5G
TCC2084-25GEA 305 USD
TCC2084-25G TCC2084-5G
Indole-6-carbonitrile ≥98.0% (by GC)
Indole-6-carbonitrile

CAS Number: 15861-36-6
MDL Number: MFCD00016732
Molecular Formula: C9H6N2
Molecular Weight: 142.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 130


Other Product Names / Synonyms: 6-Indolecarbonitrile

Formula: C₉H₆N₂
MW: 142.16 g/mol
Melting Pt: 130 °C
MDL Number: MFCD00016732
CAS Number: 15861-36-6
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a14474884ae6101459106f09d0f6b884f681ffd&vid=8a14474884ae6101459106f09d0f6b884f681ffd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR