4-(4-Chlorophenyl)morpholine ≥98.0% (by GC)

Supplier: TCI America

C2082-25G C2082-5G
TCC2082-25GEA 243 USD
TCC2082-25G TCC2082-5G
4-(4-Chlorophenyl)morpholine ≥98.0% (by GC)
4-(4-Chlorophenyl)morpholine

CAS Number: 70291-67-7
Molecular Formula: C10H12ClNO
Molecular Weight: 197.66
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 73

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR