3-Chloro-4-fluorobenzonitrile ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Chlor-3-cyan-6-fluorbenzene

C2081-25G C2081-5G
TCC2081-25GEA 173 USD
TCC2081-25G TCC2081-5G
3-Chloro-4-fluorobenzonitrile ≥98.0% (by GC)
3-Chloro-4-fluorobenzonitrile

CAS Number: 117482-84-5
MDL Number: MFCD00015431
Molecular Formula: C7H3ClFN
Molecular Weight: 155.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 135
Melting point (°C): 72

Formula: C₇H₃ClFN
MW: 155.56 g/mol
Boiling Pt: 135 °C
Melting Pt: 72 °C
MDL Number: MFCD00015431
CAS Number: 117482-84-5
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcf748c444126798b70cfec9e522bf095048f3a9&vid=dcf748c444126798b70cfec9e522bf095048f3a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR