2-Chloro-5-fluorobenzonitrile ≥98.0% (by GC)

Supplier: TCI America

C2080-1G C2080-5G
TCC2080-1GEA 43 USD
TCC2080-1G TCC2080-5G
2-Chloro-5-fluorobenzonitrile ≥98.0% (by GC)
2-Chloro-5-fluorobenzonitrile

CAS Number: 57381-56-3
MDL Number: MFCD03094168
Molecular Formula: C7H3ClFN
Molecular Weight: 155.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 84

Formula: C₇H₃ClFN
MW: 155.56 g/mol
Melting Pt: 84 °C
MDL Number: MFCD03094168
CAS Number: 57381-56-3
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b74ee43c8c01c573f20a833cd6df4eaeecf4bbd4&vid=b74ee43c8c01c573f20a833cd6df4eaeecf4bbd4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR